Pantoprazole sodium salt hydrate
Fornitore: Thermo Scientific
|
Attenzione
|
460870010
460870050
ACRO460870010EA
44.1
EUR
ACRO460870010
ACRO460870050
Pantoprazole sodium salt hydrate
Pantoprazole sodium salt hydrate
Formula:
C₁₆H₁₄F₂N₃NaO₄S·xH₂O Punto di fusione: 151…155 °C Temperatura di stoccaggio: Congelamento |
MDL Number:
MFCD08704580 CAS Number: 718635-09-7 |
Risultati del test di specificazione
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Water | 4 to 6 % |
Solubility | (1 % in water) Clear solution |
Titration with HClO4 | 98 to 102 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...