Mexiletine hydrochloride
Fornitore: Thermo Scientific
|
Attenzione
|
Synonyms:
1-(2,6-Dimethylphenoxy)propan-2-amine hydrochloride, 1-(2,6-Dimethylphenoxy)-2-propylamine Hydrochloride, Mexiletine HCl
372910050
372910250
372911000
ACRO372910050EA
29.1
EUR
ACRO372910050
ACRO372910250
ACRO372911000
Mexiletine hydrochloride
Mexiletine hydrochloride
Formula:
C₁₁H₁₇NO Peso molecolare: 215,72 g/mol Punto di fusione: 202 °C Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00216024 CAS Number: 5370-01-4 |
Risultati del test di specificazione
Appearance | White to off-white Powder |
Infrared spectrum | Conforms |
Melting point | 200 to 205 °C |
Loss on drying | ≤0.5 % |
HPLC | ≥97.5 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...