Glycogen (from beef liver)
Fornitore: Thermo Fisher Scientific
Formula:
C₂₄H₄₂O₂₁ Temperatura di stoccaggio: Refrigerata |
MDL Number:
MFCD00081547 CAS Number: 9005-79-2 |
Risultati del test di specificazione
Appearance | White to beige Powder |
Infrared spectrum | Conforms |
Loss on drying | ≤15 % (105°C, 2 hr) |
Specific optical rotation | +180° to +200° (20°C, 589 nm) (0.1 % in water) (on dry substance) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...