Stampa…

4,4′-Azo-bis(4-cyanovaleric acid), (∼18% H₂O) 98%

Fornitore: Thermo Fisher Scientific
Pericolo

H59611.14 H59611.22
H59611.14EA 86.8 EUR
H59611.14 H59611.22
4,4′-Azo-bis(4-cyanovaleric acid), (∼18% H₂O) 98%
4,4′-Azo-bis(4-cyanovaleric acid)

4,4'-Azobis(4-cyanovaleric acid) is used as a free radical imitator in polymer synthesis such as polyvinyl chloride, polyacrylonitrile, polyvinyl alcohol, and synthetic fibres.


  • Free radical imitator
  • Blowing agent for plastics and elastomers


It is utilised for both heterogeneous and homogenous free-radical polymerisations and as an initiator in reversible addition-fragmentation chain transfer polymerisation (RAFT). It is also used as a blowing agent for plastics and elastomers.


Attenzione: Light sensitive. Incompatible with strong oxidising agents.

Formula: C₁₂H₁₆N₄O₄
Punto di fusione: 118…125 °C (dec.)
Temperatura di stoccaggio: Refrigerata
MDL Number: MFCD00002799
CAS Number: 2638-94-0
EINECS: 220-135-0
UN: 1325
ADR: 4.1,II

Visualizza i codici

Risultati del test di specificazione

Solubility Soluble in water
Sensitivity Light sensitive


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR