Stampa…

Organic reference standards, Tenoxicam standard, BP

Fornitore: LGC Standards PROMOCHEM

BP708 BP709
PROOBP708EA 298 EUR
PROOBP708 PROOBP709
Organic reference standards, Tenoxicam standard, BP
Standard Standard organici

Visualizza i codici

Specifiche

Descrizione Tenoxicam assay standard Tenoxicam degradation impurity standard
Environmentally Preferable
CAS No. 59804-37-4 59804-37-4
Formula C₁₃H₁₁N₃O₄S₂ C₁₃H₁₁N₃O₄S₂

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR