Stampa…
Anti-vibration gloves, ActivArmr® 07-112
Fornitore: Ansell
07-112/10 07-112/9 07-112/8
112-4235EA 77.4 EUR
112-4235 112-4237 112-4236
Anti-vibration gloves, ActivArmr® 07-112
Guanti
The VibraGuard® gloves feature a nitrile impregnated coating on a cotton interlock/Gelfôm™ liner. They provide resistance to cuts, punctures and abrasion, as well as to grease and oil.
  • Ideal for tasks involving vibrations at frequency levels greater than 300 Hz
  • Engineered to reduce the vibration absorbed by workers' palm and fingers
  • Help to protect from Hand-Arm Vibration Syndrome
Ideal for hand and pedestal grinders, vibrating poker, road drills, and applications including chipping hammers, heavy-duty hammers and impact rammers.
Vibration attenuation EN 10819: TRM (Transmission Ratio Medium frequency range) = 0,90/TRH (Transmission Ratio High frequency range) = 0,52
Certificazioni: EN 388:2016 (3221B); Cat. II