Stampa…

Nickel chromium iron, in polvere -325 mesh

Fornitore: Thermo Scientific
Attenzione

44888.30 44888.A1
44888.30EA 200 EUR
44888.30 44888.A1
Nickel chromium iron, in polvere -325 mesh
Nickel chromium iron

Molecular Formula: Ni:Cr:Fe; 58:27-31:7-11 wt% UN No.: 3089 MDL No.: MFCD01741927

Formula: Ni:Cr:Fe;58:27-31:7-11%
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01741927
UN: 3089
ADR: 4.1,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR