Stampa…

Methacholine chloride ≥98%, white powder

Fornitore: MP Biomedicals
Attenzione

Synonyms: Mecholyl Chloride, O-Acetyl-beta-methylcholine Chloride, Acetyl-β-methylcholine chloride

0219023180 0219023105 0219023110 0219023125
ICNA0219023180EA 381 EUR
ICNA0219023180 ICNA0219023105 ICNA0219023110 ICNA0219023125
Methacholine chloride ≥98%, white powder
Methacholine chloride

Methacholine chloride is a cholinergic agonist and parasympathomimetic bronchoconstrictor. A metabolically stable analog of acetylcholine. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.
Methacholine is primarily used to diagnose bronchial hyperreactivity, which is the hallmark of asthma and also occurs in chronic obstructive pulmonary disease. Acetyl-β-methylcholine (methacholine), an analog of acetylcholine, is use as a receptor agonist to study processes such as airway hyperresponsiveness associated with asthma.
Metabolically stable analog of acetylcholine; muscarinic agonist. Its ability to constrict airway smooth muscle is used to assess bronchial reactivity in the laboratory and clinically.


  • Presentation: White Powder
  • Soluble in water (100 mg/mL-clear, colorless solution), alcohol, chloroform; Insoluble in ether.
  • -20°C, desiccate


Storage temperature.: +4 °C

Formula: C₈H₁₈ClNO₂
Peso molecolare: 195,69 g/mol
Punto di fusione: 164…174 °C
Temperatura di stoccaggio: Congelamento
MDL Number: MFCD00011817
CAS Number: 62-51-1
EINECS: 200-537-2

Visualizza i codici

Risultati del test di specificazione

Identity Test Passes
Purity ≥98%
FTIR Conforms to Standard

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR