Stampa…
Lecithin
Fornitore: MP Biomedicals
0210214801 0210214805
ICNA0210214801EA 55.3 EUR
ICNA0210214801 ICNA0210214805
Lecithin
Lecithin
Lecithin is a good source of choline, which may help with brain function and growth. Used as an emulsifier and solubilizer for drugs in an aqueous solution, also as a vehicle for liposomes. Lecithin is a phosphatidylcholine of soybean origin.
Storage temperature: -20 °C
MDL Number: MFCD00147406
CAS Number: 8002-43-5
Merck Index: 12,05452