Stampa…
Flame retardant overalls, MICROGARD® FR
Fornitore: Ansell
BL95S-00111-04 BL95S-00111-05 BL95S-00111-06 BL95S-00111-07 BL95S-00111-03
ANSEBL95S-00111-04EA 622 EUR
ANSEBL95S-00111-04 ANSEBL95S-00111-05 ANSEBL95S-00111-06 ANSEBL95S-00111-07 ANSEBL95S-00111-03
Flame retardant overalls, MICROGARD® FR
Abbigliamento da lavoro
MICROGARD® FR is a flame retardant material designed to be worn over woven thermal protective garments such as NOMEX® or PYROVATEX®, offering protection from particulates and pressurised liquid spray without compromising worker protection in the event of a flash fire.
  • Flame retardant treated Sontara/wood pulp/polyester fabric with good barrier to particulates and low level liquid spray
  • Air and water vapour permeable (breathable) to help reduce the risk of heat stress
  • Optimised body fit improves wearer comfort and safety
  • Elasticated 2-piece hood, wrists, waist and ankles
  • External overlocked, stitched seams
MICROGARD® FR offers wearers protection from liquid chemicals to EN Type 6 and particulates to EN Type 5, and peace of mind to workers in potentially explosive/flammable environments. To decrease the risk of burn injury wear over thermal protective workwear.
Typical applications include: Oil and petrochemicals, petroleum distribution and processing, utilities.
Certificazioni: Type 5/6, EN ISO 14116 (index 1/0/0), EN 1073-2, EN 1149-5; Cat. III
Attenzione: MICROGARD® FR should never be worn in isolation for flame retardant protection. Always wear over the top of garments which achieve EN ISO 14116 Index 2 or above.