Stampa…
Label printer, BMP51 and BMP53 application kits
Fornitore: Brady
710938 710929 710931 710940
BRDY710938EA 635 EUR
BRDY710938 BRDY710929 BRDY710931 BRDY710940
Label printer, BMP51 and BMP53 application kits
Stampanti Label Printers
These kits offer a printer with software and a selection of labels to accurately identify samples in the laboratory. Both the BMP51 and BMP53 are portable, single colour, thermal printers with 300 dpi print head, and with very similar specifications except the BMP51 has stand-alone capability and the BMP53 does not.
  • USB, Bluetooth and optional Ethernet, Wi-Fi connectivity
  • Print speed 25,4 mm/second
BMP51 Laboratory identification kit has qwerty keyboard for on the spot sample identification. The top loading label cartridge contains both the printer ribbon and labels and can be exchanged in seconds.
Informazioni per l'ordine: Electrical kit includes BMP51 or BMP53 printer with hard case, USB cable, magnet, rechargeable Li-on battery and AC adapter with power cord, LabelMark™ 5 labelling software; automation and electrical labels, control panel and electrical panel labels, cable labels. Teledatacom kit includes BMP51 or BMP53 printer with hard case, USB cable, magnet, rechargeable Li-on battery and AC adapter with power cord, LabelMark™ 5 labelling software, wire and cable labels, punch block and patch panel labels, control panel and electrical panel labels.