Stampa…
Venting filters, SteriVENT™, Whatman™
Fornitore: Whatman products (Cytiva)
2103 2107 2108
WHAT2103EA 134 EUR
WHAT2103 WHAT2107 WHAT2108
Venting filters, SteriVENT™, Whatman™
Filtri Filtri di ventilazione
SteriVENT™ venting filters with an integral hydrophobic PTFE membrane and PP housing are well suited for sterile venting of vessels and tanks during draining and filling. These filters are capable of allowing clean air to pass into the vessel while preventing microbiological contamination.
  • Industrial air filters for sterile venting
  • Can be autoclaved 50 times without loss of performance; compatible with ethylene oxide (EtO)
  • Integrity can be tested with the water break through (WBT) test
  • Bidirectional flow direction
  • Heat fused sealing
  • Vent on inlet
Visualizza i codici
Specifiche
Environmentally Preferable
Ø 36 mm 150 mm 150 mm
Tipo di alloggiamento SteriVENT™ 36 capsule SteriVENT™ 150 capsule SteriVENT™ 150 capsule
Pressione max. 2 bar 2 bar 2 bar
Ingresso/uscita 3/8" to 1/2" stepped hose barb 3/8" to 1/2" stepped hose barb 1 1/2" sanitary/1 1/2" sanitary