Stampa…
Overalls, 4500
Fornitore: 3M
4500WS 4500WM 4500WXL 4500W2XL 4500W3XL 4500W4XL 4500WL
MMMA4500WSEA 169 EUR
MMMA4500WS MMMA4500WM MMMA4500WXL MMMA4500W2XL MMMA4500W3XL MMMA4500W4XL MMMA4500WL
Overalls, 4500
Indumenti usa e getta
This simple overall made of durable PP fabric is suitable for non hazardous applications where hygiene and cleanliness are important.
  • Excellent economic solution to keep the wearer clean or dry
  • Breathable and light duty protection
  • Two-way zip with storm flap
  • Elasticated 2-panel hood, cuffs, ankles and waist
Typical applications include: Domestic DIY use, light duty industrial cleaning and maintenance applications.
Weight: 45 g/m²
Certificazioni: Cat. I
Visualizza i codici
Specifiche
Modello 4500
Environmentally Preferable
Tessuto PP
Genere
Applicazioni per abiti General Purpose,Laboratory,Industrial
Peso 45 g/m²