Stampa…

Anti-NEFH & NEFL Mouse Monoclonal Antibody (CF488A) [clone: RT-97 NR-4]

Fornitore: Biotium
BNC881201-500 BNC881201-100
BNC881201-500EA 560 EUR
BNC881201-500 BNC881201-100
Anti-NEFH & NEFL Mouse Monoclonal Antibody (CF488A) [clone: RT-97 NR-4]
Anticorpi
This MAb reacts with a 200 kDa and 68 kDa protein, identified as heavy and light sub-units of neurofilaments (NF-H & NF-L). Neurofilaments make up the main structural elements of axons and dendrites and are found in neurons, peripheral nerves, and sympathetic ganglion cells. Neurofilaments consist of three major subunits with molecular weights of 68 kDa (NF-L), 160 kDa (NF-M) and 200 kDa (NF-H). Anti-neurofilament stains a number of neural, neuroendocrine, and endocrine tumors. Neuromas, ganglioneuromas, gangliogliomas, ganglioneuroblastomas, and neuroblastomas stain positively for anti-neurofilament. Neurofilaments are also present in paragangliomas as well as adrenal and extra-adrenal pheochromocytomas. Carcinoids, neuroendocrine carcinomas of the skin, and oat cell carcinomas of the lung also express neurofilament.

Type: Primary
Antigen: NEFH & NEFL
Clonality: Monoclonal
Clone: RT-97 NR-4
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Chicken, Human, Mouse, Pig, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR