Stampa…
Anti-EGFR Mouse Monoclonal Antibody (CF488A) [clone: GFR1195 31G7]
Fornitore: Biotium
BNC881200-500 BNC881200-100
BNC881200-500EA 560 EUR
BNC881200-500 BNC881200-100
Anti-EGFR Mouse Monoclonal Antibody (CF488A) [clone: GFR1195 31G7]
Anticorpi
This MAb recognizes a protein of 170 kDa, identified as EGFR. EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin. EGFR is overexpressed in tumors of breast, brain, bladder, lung, gastric, head & neck, esophagus, cervix, vulva, ovary, and endometrium. It is predominantly present in squamous cell carcinomas.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: GFR1195 31G7
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Human