Stampa…

Anti-IDH1 Mouse Monoclonal Antibody (Purified) [clone: IDH1/1152]

Fornitore: Biotium

BNUB1152-500 BNUB1152-100
BNUB1152-500EA 483 EUR
BNUB1152-500 BNUB1152-100
Anti-IDH1 Mouse Monoclonal Antibody (Purified) [clone: IDH1/1152]
Anticorpi
This antibody recognizes a 45 kDa protein, which is identified as isocitrate dehydrogenase (IDH1). It belongs to the isocitrate and isopropylmalate dehydrogenases family. IDH1 catalyzes the third step of the citric acid cycle, which involves the oxidative decarboxylation of isocitrate, formingalpha-ketoglutarate and CO2 in a two-step reaction. The first step involves the oxidation of isocitrate to the intermediate oxalosuccinate, while the second step involves the production ofalpha-ketoglutarate. During this process, either NADH or NADPH is produced along with CO2. Recently, an inactivating mutation of IDH1 has been implicated in glioblastoma. IDH1 appears to function as a tumor suppressor that, when mutationally inactivated, contributes to tumorigenesis in part through induction of the HIF-1 pathway.

Type: Primary
Antigen: IDH1
Clonality: Monoclonal
Clone: IDH1/1152
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR