Stampa…
Anti-TFRC Mouse Monoclonal Antibody (CF568) [clone: TFRC/1149]
Fornitore: Biotium
BNC681149-500 BNC681149-100
BNC681149-500EA 560 EUR
BNC681149-500 BNC681149-100
Anti-TFRC Mouse Monoclonal Antibody (CF568) [clone: TFRC/1149]
Anticorpi
This antibody recognizes a ~90-95 kDa protein which is identified as cell surface transferrin receptor (CD71), a disulfide-bonded homodimeric glycoprotein of 180-190 kDa. This MAb is highly specific to CD71 and shows no cross-reaction with other related proteins. Ligand for transferrin receptor is the serum iron transport protein, transferrin. This receptor is broadly distributed in carcinomas, sarcomas, leukemias, and lymphomas. CD71/Transferrin receptor has been reported to be associated with cell proliferation in both normal and neoplastic tissues and useful in predicting clinical behavior or response to therapy in a number of malignancies including breast cancer.
Type: Primary
Antigen: TFRC
Clonality: Monoclonal
Clone: TFRC/1149
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human