Stampa…
Anti-PTPRC Mouse Monoclonal Antibody (CF640R) [clone: PTPRC/1132]
Fornitore: Biotium
BNC401132-500 BNC401132-100
BNC401132-500EA 560 EUR
BNC401132-500 BNC401132-100
Anti-PTPRC Mouse Monoclonal Antibody (CF640R) [clone: PTPRC/1132]
Anticorpi
CD45R, also designated CD45 and PTPRC, is identified as a transmembrane glycoprotein, broadly expressed among hematopoietic cells. Multiple isoforms of CD45R are distributed throughout the immune system. These isoforms arise because of alternative splicing of exons 4, 5, and 6. The corresponding protein domains are characterized by the binding of monoclonal antibodies specific for CD45RA (exon 4), CD45RB (exon 5), CD45RC (exon 6) and CD45RO (exons 4 to 6 spliced out). The variation in these isoforms is localized to the extracellular domain of CD45R, while the intracellular domain isMore Product Information