Stampa…

Anti-Cytokeratin Mouse Monoclonal Antibody [clone: KRTL/1077 KRTH/1076]

Fornitore: Biotium
BNUB1100-500 BNUB1100-100
BNUB1100-500EA 511 EUR
BNUB1100-500 BNUB1100-100
Anti-Cytokeratin Mouse Monoclonal Antibody [clone: KRTL/1077 KRTH/1076]
Anticorpi
Twenty human keratins are resolved with two-dimensional gel electrophoresis into acidic (pI 6.0) subfamilies. This antibody cocktail recognizes acidic (Type I or LMW) and basic (Type II or HMW) cytokeratins, which include CK1, CK3, CK4, CK5, CK6, CK8, CK10, CK14, CK15, CK16, and CK19. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis. KRTL/KRTH is a broad spectrum anti pan-cytokeratin antibody cocktail, which differentiates epithelial tumors from non-epithelial tumors e.g. squamous vs. adenocarcinoma of the lung, liver carcinoma, breast cancer, and esophageal cancer. It has been used to characterize the source of various neoplasms and to study the distribution of cytokeratin containing cells in epithelia during normal development and during the development of epithelial neoplasms. This antibody stains cytokeratins present in normal and abnormal human tissues and has shown high sensitivity in the recognition of epithelial cells and carcinomas.

  • Purified

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: KRTL/1077 KRTH/1076
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR