Stampa…
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Fornitore: Novus Biologicals
NBP1-50631 NBP1-50631SS
NOVUNBP1-50631EA 554 EUR
NOVUNBP1-50631 NOVUNBP1-50631SS
Anti-LAMTOR3 Rabbit Polyclonal Antibody
Anticorpi
Rabbit Polyclonal MAP2K1IP1/MAPKSP1 Antibody. Tested Applications: Western Blot, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunohistochemistry-Paraffin. Tested Reactivity: Human, Mouse.
Recommended Dilutions: Immunohistochemistry (Parafin): 1:100
Type: Primary
Antigen: LAMTOR3
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human