Stampa…

Anti-IFNA1 Mouse Monoclonal Antibody (CF405S) [clone: 2.52]

Fornitore: Biotium
BNC040998-500 BNC040998-100
BNC040998-500EA 560 EUR
BNC040998-500 BNC040998-100
Anti-IFNA1 Mouse Monoclonal Antibody (CF405S) [clone: 2.52]
Anticorpi
This MAb is specific for human interferon alpha 1 and does not cross react with human interferon alpha 2. Interferons are widely used therapeutic agents because of their anti-tumor and anti-viral effects and because of their modulatory effects on the immune system. These cytokines produce their effects by binding to the Type 1 Interferon- & Receptor (IFNAR1). Down regulation of this receptor plays a key role in determining the magnitude and duration of cytokine signaling. This down regulation is influenced by phosphorylation of Serine 535 and 539 in the IFNAR1.

Type: Primary
Antigen: IFNA1
Clonality: Monoclonal
Clone: 2.52
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR