Stampa…
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF640R) [clone: KRT8/803 KRT18/835]
Fornitore: Biotium
BNC400979-500 BNC400979-100
BNC400979-500EA 560 EUR
BNC400979-500 BNC400979-100
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF640R) [clone: KRT8/803 KRT18/835]
Anticorpi
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 1More Product Information