Stampa…

Anti-CD47 Mouse Monoclonal Antibody (CF640R) [clone: IAP/964]

Fornitore: Biotium

BNC400964-500 BNC400964-100
BNC400964-500EA 530 EUR
BNC400964-500 BNC400964-100
Anti-CD47 Mouse Monoclonal Antibody (CF640R) [clone: IAP/964]
Anticorpi
This antibody reacts with Ig domain of CD47 protein. CD47, originally named integrin-associated protein (IAP), is a 50 kDa protein containing five membrane-spanning sequences and a short cytoplasmic tail. CD47 plays a role in both cell adhesion by acting as an adhesion receptor for THBS1 on platelets, and in the modulation of integrins. It is important in memory formation and synaptic plasticity in the hippocampus. CD47 may play a role in membrane transport and/or integrin dependent signal transduction.

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: IAP/964
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR