Stampa…

Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: MDA/929]

Fornitore: Biotium
BNC400929-500 BNC400929-100
BNC400929-500EA 560 EUR
BNC400929-500 BNC400929-100
Anti-CD14 Mouse Monoclonal Antibody (CF640R) [clone: MDA/929]
Anticorpi
Recognizes a protein of 55 kDa, identified as CD14 (also known lipopolysaccharide receptor). CD14 is expressed strongly on monocytes and macrophage and weakly on the surface of neutrophils. CD14 is anchored to cells by linkage to glycosylphosphatidylinositol (GPI) and functions as a high affinity receptor for complexes of LPS and LPS binding protein (LBP). Soluble CD14, also binding to LPS, acts at physiological concentration as an LPS agonist and has, at higher concentrations, an LPS antagonizing effect in cell activation.

Type: Primary
Antigen: CD14
Clonality: Monoclonal
Clone: MDA/929
Conjugation: CF640R
Epitope:
Host: Mouse
Reactivity: Human
Isotype: IgG1, kappa
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR