Stampa…

Anti-MYH11 Mouse Monoclonal Antibody (CF488A) [clone: MYH11/923]

Fornitore: Biotium
BNC880923-500 BNC880923-100
BNC880923-500EA 560 EUR
BNC880923-500 BNC880923-100
Anti-MYH11 Mouse Monoclonal Antibody (CF488A) [clone: MYH11/923]
Anticorpi
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breastMore Product Information
Visualizza i codici

Specifiche

Simbolo dell'antigene MYH11
Environmentally Preferable
Tipo di anticorpi Myosin Smooth Muscle Heavy Chain
Sinomini antigene SMMHC|AAT4|Myosin heavy chain 11 smooth muscle|SM2|Smooth muscle myosin heavy chain 11|Myosin-11|Myosin heavy chain 11 (MYH11)|SM1|SMHC
Tipo anticorpo Primary
Clonalità Monoclonale
Coniugazione CF® 488A
Clone Clone MYH11/923
Reattività Human,Rat
Ospite Mouse
ID gene 460109
Isotipo IgG1 kappa
ImmunoChimica Yes
ImmunoFluorescenza Yes
Concentrazione 0,1 mg/ml
Tampone di conservazione PBS/0,1% BSA/0,05% azide

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR