Stampa…
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: MYH11/923]
Fornitore: Biotium
BNCB0923-500 BNCB0923-100
BNCB0923-500EA 560 EUR
BNCB0923-500 BNCB0923-100
Anti-MYH11 Mouse Monoclonal Antibody (Biotin) [clone: MYH11/923]
Anticorpi
Smooth muscle myosin heavy chain (SM-MHC) is a cytoplasmic structural protein, which is a major component of the contractile apparatus in smooth muscle cells. Expression of smooth muscle myosin is developmentally regulated, appearing early in smooth muscle development, and is specific for smooth muscle development. Two isoforms of smooth muscle myosin heavy chain have been identified, designated MHC-1 and MHC-2. The antibody may be useful for the study of breast tumors as the presence of an intact layer of myoepithelial cells is an important feature, which may distinguish benign breastMore Product Information