Stampa…

Anti-TYR Mouse Monoclonal Antibody (CF594) [clone: T311 OCA1/812]

Fornitore: Biotium
BNC940908-500 BNC940908-100
BNC940908-500EA 560 EUR
BNC940908-500 BNC940908-100
Anti-TYR Mouse Monoclonal Antibody (CF594) [clone: T311 OCA1/812]
Anticorpi
Recognizes a cluster of proteins between 70-80 kDa, identified as tyrosinase. Occasionally a minor band at 55 kDa is also detected. This MAb shows no cross-reaction with MAGE-1 and tyrosinase-related protein 1, TRP-1/gp75. Tyrosinase is a copper-containing metalloglycoprotein that catalyzes several steps in the melanin pigment biosynthetic pathway; the hydroxylation of tyrosine to L-3,4-dihydroxy-phenylalanine (dopa), and the subsequent oxidation of dopa to dopaquinone. Mutations of the tyrosinase gene occur in various forms of albinism. Tyrosinase is one of the targets for cytotoxic T-cell recognition in melanoma patients. Staining of melanomas with this MAb shows tyrosinase in melanotic as well as amelanotic variants. This MAb is a useful marker for melanocytes and melanomas.

Type: Primary
Antigen: TYR
Clonality: Monoclonal
Clone: T311 OCA1/812
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR