Stampa…
Anti-MUC6 Mouse Monoclonal Antibody (Purified) [clone: CLH5]
Fornitore: Biotium
BNUB0891-500 BNUB0891-100
BNUB0891-500EA 511 EUR
BNUB0891-500 BNUB0891-100
Anti-MUC6 Mouse Monoclonal Antibody (Purified) [clone: CLH5]
Anticorpi
The MUC6 gastric mucin is a secreted glycoprotein that plays an essential role in epithelial cyto-protection from acid, proteases, pathogenic microorganisms, and mechanical trauma in the gastrointestinal tract. Mucin 6 expression is highest in the stomach and gall bladder, with lower expression in the terminal ileum and right colon. In gastric cancer, Mucin 6 has an altered expression. In normal stomach, Mucin 6 is associated with Lewis type 2; Mucin 6 is also expressed in gastric metaplasia, duodenum and pancreas. Mucin 6 is a secretory mucin, located in the deeper mucosal folds of human gall bladder, and its expression is altered with increasing degrees of inflammation.
Type: Primary
Antigen: MUC6
Clonality: Monoclonal
Clone: CLH5
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human