Stampa…

Anti-CALD1 Mouse Monoclonal Antibody (CF647) [clone: h-CALD]

Fornitore: Biotium
BNC470819-500 BNC470819-100
BNC470819-500EA 560 EUR
BNC470819-500 BNC470819-100
Anti-CALD1 Mouse Monoclonal Antibody (CF647) [clone: h-CALD]
Anticorpi
Recognizes a protein of 150 kDa, which is identified as the high molecular weight variant of Caldesmon. Two closely related variants of human caldesmon have been identified which are different in their electrophoretic mobility and cellular distribution. The h-caldesmon variant (120-150 kDa) is predominantly expressed in smooth muscle whereas l-caldesmon (70-80 kDa) is found in non- muscle tissue and cells. Neither of the two variants has been detected in skeletal muscle. This MAb recognizes only the 150 kDa variant (h-caldesmon) in Western blots of human aortic media extracts and is unreactive with fibroblast extracts from cultivated human foreskin. Caldesmon is a developmentally regulated protein involved in smooth muscle and non-muscle contraction.

Type: Primary
Antigen: CALD1
Clonality: Monoclonal
Clone: h-CALD
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR