Stampa…
Anti-BCL2 Mouse Monoclonal Antibody (CF594) [clone: BCL2/796]
Fornitore: Biotium
BNC940796-500 BNC940796-100
BNC940796-500EA 560 EUR
BNC940796-500 BNC940796-100
Anti-BCL2 Mouse Monoclonal Antibody (CF594) [clone: BCL2/796]
Anticorpi
This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits the programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicularMore Product Information