Stampa…
Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: ERB2/776]
Fornitore: Biotium
BNC400776-500 BNC400776-100
BNC400776-500EA 560 EUR
BNC400776-500 BNC400776-100
Anti-ERBB2 Mouse Monoclonal Antibody (CF640R) [clone: ERB2/776]
Anticorpi
This MAb is specific to c-erbB-2/HER-2 and shows minimal cross-reaction with other members of the family. C-erbB-2/HER-2 is a member of the EGFR family. Receptors of this family are located on the plasma membrane and consist of an extracellular ligand-binding domain that is connected to a large intracellular domain by a single transmembrane sequence. c-erbB-2/HER-2 protein is over-expressed in a variety of carcinomas especially those of breast and ovary.
Type: Primary
Antigen: ERBB2
Clonality: Monoclonal
Clone: ERB2/776
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human