Stampa…

Anti-CDKN1B Mouse Monoclonal Antibody (Purified) [clone: KIP1/769]

Fornitore: Biotium
BNUB0769-500 BNUB0769-100
BNUB0769-500EA 511 EUR
BNUB0769-500 BNUB0769-100
Anti-CDKN1B Mouse Monoclonal Antibody (Purified) [clone: KIP1/769]
Anticorpi
This MAb recognizes a 27 kDa protein, identified as the p27Kip1, a cell cycle regulatory mitotic inhibitor. It is highly specific and shows no cross-reaction with other related mitotic inhibitors. p27Kip1 functions as a negative regulator of G1 progression and has been proposed to function as a possible mediator of TGF- induced G1 arrest. p27Kip1 is a candidate tumor suppressor gene. This MAb is excellent for staining of formalin-fixed tissues.

Type: Primary
Antigen: CDKN1B
Clonality: Monoclonal
Clone: KIP1/769
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR