Stampa…
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: K72.7]
Fornitore: Biotium
BNC400757-500 BNC400757-100
BNC400757-500EA 560 EUR
BNC400757-500 BNC400757-100
Anti-KRT7 Mouse Monoclonal Antibody (CF640R) [clone: K72.7]
Anticorpi
This antibody recognizes an intermediate filament protein (IFP) of 55 kDa, which is identified as cytokeratin 7. This MAb is highly specific to cytokeratin 7 and shows no cross-reaction with other IFPs. Cytokeratin 7 is a basic cytokeratin, which is found in most glandular and transitional epithelia but not in the stratified squamous epithelia. Keratin 7 is expressed in the epithelial cells of ovary, lung, and breast but not of colon, prostate, or gastrointestinal tract. This MAb is highly useful in distinguishing ovarian carcinomas (keratin 7 ) from colon carcinomas (keratin 7-).
Type: Primary
Antigen: KRT7
Clonality: Monoclonal
Clone: K72.7
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human