Stampa…

Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF647) [clone: K8.8 DC10]

Fornitore: Biotium
BNC470691-500 BNC470691-100
BNC470691-500EA 560 EUR
BNC470691-500 BNC470691-100
Anti-KRT8 & KRT18 Mouse Monoclonal Antibody (CF647) [clone: K8.8 DC10]
Anticorpi
Cytokeratin 8 (CK8) belongs to the type II (or B or basic) subfamily of high molecular weight cytokeratins and exists in combination with cytokeratin 18 (CK18). This MAb cocktail recognizes all simple epithelia including glandular epithelium, for example thyroid, female breast, gastrointestinal tract, respiratory tract, and urogenital tract including transitional epithelium. All adenocarcinomas and most squamous carcinomas are positive but keratinizing squamous carcinomas are usually negative. This antibody is useful in demonstrating the presence of Paget cells; there is very little keratin 18 in the normal epidermis so only Paget cells are stained.Immunohistochemical staining with this MAb is indistinguishable from that obtained with monoclonal antibody 5D3.

Type: Primary
Antigen: KRT8 & KRT18
Clonality: Monoclonal
Clone: K8.8 DC10
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR