Stampa…

Anti-Il6 (Mouse) Rat Monoclonal Antibody (Purified) [clone: MP5-20F3]

Fornitore: Biotium
BNUB0663-500 BNUB0663-100
BNUB0663-500EA 511 EUR
BNUB0663-500 BNUB0663-100
Anti-Il6 (Mouse) Rat Monoclonal Antibody (Purified) [clone: MP5-20F3]
Anticorpi
IL-6 is a potent lymphoid cell growth factor that stimulates the growth and survivability of certain B-cells and T-cells. IL-6 plays a role in host defense, acute phase reactions, immune responses, and hematopoiesis. IL-6 is expressed by T-cells, B-cells, monocytes, fibroblasts, hepatocytes, endothelial cells and keratinocytes.

Type: Primary
Antigen: Il6 (Mouse)
Clonality: Monoclonal
Clone: Clone MP5-20F3
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1 kappa
Reactivity: Mouse
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR