Stampa…
Anti-TNF Mouse Monoclonal Antibody (CF647) [clone: TNF655]
Fornitore: Biotium
BNC470655-500 BNC470655-100
BNC470655-500EA 560 EUR
BNC470655-500 BNC470655-100
Anti-TNF Mouse Monoclonal Antibody (CF647) [clone: TNF655]
Anticorpi
This MAb recognizes human 17-26 kDa protein, which is identified as cytokine TNF-alpha (Tumor Necrosis Factor-alpha). TNF-alpha can be expressed as a 17 kDa free molecule, or as a 26 kDa membrane protein. TNF-alpha is a protein secreted by lipopolysaccharide-stimulated macrophages, and causes tumor necrosis when injected into tumor bearing mice. TNF alpha is believed to mediate pathogenic shock and tissue injury associated with endotoxemia. TNF alpha exists as a multimer of two, three, or five non-covalently linked units, but shows a single 17 kDa band following SDS PAGE under non-reducingMore Product Information