Stampa…

Anti-EGFR Mouse Monoclonal Antibody (CF594) [clone: B1D8]

Fornitore: Biotium
BNC940643-500 BNC940643-100
BNC940643-500EA 560 EUR
BNC940643-500 BNC940643-100
Anti-EGFR Mouse Monoclonal Antibody (CF594) [clone: B1D8]
Anticorpi
This antibody reacts with the extracellular domain of EGFR and blocks the EGF/TGF alpha induced activation. It also blocks tumor growth in vivo. It is excellent for purification of EGFR. EGFR is type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin and epiregulin.

Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: B1D8
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR