Stampa…
Anti-EGFR Mouse Monoclonal Antibody (Purified) [clone: H9B4]
Fornitore: Biotium
BNUB0642-500 BNUB0642-100
BNUB0642-500EA 511 EUR
BNUB0642-500 BNUB0642-100
Anti-EGFR Mouse Monoclonal Antibody (Purified) [clone: H9B4]
Anticorpi
This MAb reacts with a cytoplasmic domain of EGFR. EGFR is a type I receptor tyrosine kinase with sequence homology to erbB-1, -2, -3 -4 or HER-1, -2, -3 -4. It binds to Epidermal Growth Factor (EGF), Transforming Growth Factor-a (TGF-a), Heparin-binding EGF (HB-EGF), amphiregulin, βcellulin & epiregulin.
Type: Primary
Antigen: EGFR
Clonality: Monoclonal
Clone: H9B4
Conjugation: Purified
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human