Stampa…

Anti-TMEM16A Mouse Monoclonal Antibody (CF647) [clone: DG1/447]

Fornitore: Biotium
BNC470447-500 BNC470447-100
BNC470447-500EA 560 EUR
BNC470447-500 BNC470447-100
Anti-TMEM16A Mouse Monoclonal Antibody (CF647) [clone: DG1/447]
Anticorpi
Expression of DOG-1 protein is elevated in the gastrointestinal stromal tumors (GISTs), c-kit signaling-driven mesenchymal tumors of the GI tract. DOG-1 is rarely expressed in other soft tissue tumors, which, due to appearance, may be difficult to diagnose. Immunoreactivity for DOG-1 has been reported in 97.8 percent of scorable GISTs, including all c-kit negative GISTs. Overexpression of DOG-1 has been suggested to aid in the identification of GISTs, including Platelet-Derived Growth Factor Receptor Alpha mutants that fail to express c-kit antigen. The overall sensitivity of DOG1 and c-kit in GISTs is nearly identical: 94.4% vs. 94.7%.

Type: Primary
Antigen: TMEM16A
Clonality: Monoclonal
Clone: DG1/447
Conjugation: CF647
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR