Stampa…
Anti-Cytokeratins Mouse Monoclonal Antibody (Purified) [clone: C11]
Fornitore: Biotium
BNUB0393-500 BNUB0393-100
BNUB0393-500EA 511 EUR
BNUB0393-500 BNUB0393-100
Anti-Cytokeratins Mouse Monoclonal Antibody (Purified) [clone: C11]
Anticorpi
This MAb recognizes cytokeratin 4, 5, 6, 8, 10, 13, and 18. This is a broad-spectrum antibody, which has been reported to differentiate epithelial tumors from non-epithelial tumors. Many studies have shown the usefulness of keratins as markers in cancer research and tumor diagnosis.
Type: Primary
Antigen: Cytokeratins
Clonality: Monoclonal
Clone: C11
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Bovine, Frog, Goat, Guinea Pig, Human, Marmoset, Mouse, Pig, Rat