Stampa…

Anti-CD3E Rabbit Polyclonal Antibody (Biotin) [clone: Rabbit PAb]

Fornitore: Biotium
BNCB0385-500 BNCB0385-100
BNCB0385-500EA 560 EUR
BNCB0385-500 BNCB0385-100
Anti-CD3E Rabbit Polyclonal Antibody (Biotin) [clone: Rabbit PAb]
Anticorpi
Recognizes the -chain of CD3, which consists of five different polypeptide chains (designated as γ, δ, ε, ζ, and η) with MW ranging from 16-28 kDa. The CD3 complex is closely associated at the lymphocyte cell surface with the T cell antigen receptor (TCR). Reportedly, CD3 complex is involved in signal transduction to the T cell interior following antigen recognition. The CD3 antigen is first detectable in early thymocytes and probably represents one of the earliest signs of commitment to the T cell lineage. In cortical thymocytes, CD3 is predominantly intra-cytoplasmic. However, in medullary thymocytes, it appears on the T cell surface. CD3 antigen is a highly specific marker for T cells, and is present in majority of T cell neoplasms.

Type: Primary
Antigen: CD3E
Clonality: Polyclonal
Clone: Rabbit PAb
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR