Stampa…

Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF647) [clone: Rabbit PAb]

Fornitore: Biotium

BNC470374-500 BNC470374-100
BNC470374-500EA 530 EUR
BNC470374-500 BNC470374-100
Anti-Helicobacter pylori Rabbit Polyclonal Antibody (CF647) [clone: Rabbit PAb]
Anticorpi
The spiral shaped bacteriumHelicobacter pyloriis strongly associated with inflammation of the stomach and is also implicated in the development of gastric malignancy.H. pyloriis known to cause peptic ulcers and chronic gastritis in human. It is associated with duodenal ulcers and may be involved in development of adenocarcinoma and low-grade lymphoma of mucosa associated lymphoid tissue in the stomach.This antibody stains the individual H. pylori bacterium when it presents on the surface of the epithelium or in the cytoplasm of the epithelial cells in biopsy tissue sections from the antrum and body of the stomach.

Type: Primary
Antigen: Helicobacter pylori
Clonality: Polyclonal
Clone: Rabbit PAb
Conjugation: CF647
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Helicobacter pylori
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR