Stampa…

Anti-CD1B Mouse Monoclonal Antibody (CF594) [clone: RIV12]

Fornitore: Biotium
BNC940317-500 BNC940317-100
BNC940317-500EA 560 EUR
BNC940317-500 BNC940317-100
Anti-CD1B Mouse Monoclonal Antibody (CF594) [clone: RIV12]
Anticorpi
The mouse monoclonal antibody recognizes CD1b, a 44 kDa type I glycoprotein associated with beta2-microglobulin. It is expressed on dendritic cells, Langerhans cells, thymocytes, and T acute lymphoblastic leukemia cells. The CD1 multigene family encodes five forms of the CD1 T-cell surface glycoprotein in human, designated CD1A, 1B, 1C, 1D and 1E. CD1, a type 1 membrane protein, has structural similarity to the MHC class I antigen and has been shown to present lipid antigens for recognition by T lymphocytes. Constitutive endocytosis of CD1B molecules and the differential sorting of MHC class II from lysosomes separate peptide- and lipid antigen-presenting molecules during dendritic cell maturation. CD1B is also expressed in interdigitating cells.

Type: Primary
Antigen: CD1B
Clonality: Monoclonal
Clone: RIV12
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR