Stampa…
Anti-CD48 Mouse Monoclonal Antibody (CF568) [clone: 156-4H9]
Fornitore: Biotium
BNC680307-500 BNC680307-100
BNC680307-500EA 560 EUR
BNC680307-500 BNC680307-100
Anti-CD48 Mouse Monoclonal Antibody (CF568) [clone: 156-4H9]
Anticorpi
Reacts with human CD48, a 45 kDa glycosyl phophatidyl-inositol (GPI)-anchored cell surface protein. CD48 is strongly expressed on lymphocytes and monocytes and weakly on granulocytes but is absent on platelets, fibroblasts, epithelium and endothelium. CD48 is one of marker for detecting the defects of GPI anchoring structure on the patients with paroxysmal nocturnal hemoglobulinuria (PNH) and serves as a low affinity ligand for CD2.
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: 156-4H9
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG2a, kappa
Reactivity: Human