Stampa…

Anti-NKX2-2 Mouse Monoclonal Antibody (CF594) [clone: NX2/294]

Fornitore: Biotium
BNC940294-500 BNC940294-100
BNC940294-500EA 560 EUR
BNC940294-500 BNC940294-100
Anti-NKX2-2 Mouse Monoclonal Antibody (CF594) [clone: NX2/294]
Anticorpi
Expression of NKX2.2 has been found in neuroendocrine tumors of the gut, making it a potential marker for the study of gastrointestinal neuroendocrine tumors. More recently, NKX2.2 protein was identified as a target of EWS-FLI-1, the fusion protein specific to Ewing sarcoma, and was shown to be differentially upregulated in Ewing sarcoma on the basis of array-based gene expression analysis. It acts as a valuable marker for Ewing sarcoma, with a sensitivity of 93% and a specificity of 89%, and aids in the differential diagnosis of small round cell tumors.

Type: Primary
Antigen: NKX2-2
Clonality: Monoclonal
Clone: NX2/294
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG2b kappa
Reactivity: Chicken, Human, Mouse, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR