Stampa…

Anti-MME Mouse Monoclonal Antibody (CF405S) [clone: FR4D11]

Fornitore: Biotium
BNC040248-500 BNC040248-100
BNC040248-500EA 560 EUR
BNC040248-500 BNC040248-100
Anti-MME Mouse Monoclonal Antibody (CF405S) [clone: FR4D11]
Anticorpi
Recognizes a 100 kDa glycoprotein, identified as CD10, also known as Common Acute Lymphocytic Leukemia Antigen (CALLA). It is a cell surface enzyme with neutral metalloendopeptidase activity, which inactivates a variety of biologically active peptides. CD10 is expressed on the cells of lymphoblastic, Burkitt's, and follicular germinal center lymphomas, and on cells from patients with chronic myelocytic leukemia (CML). It is also expressed on the surface of normal early lymphoid progenitor cells, immature B cells within adult bone marrow and germinal center B cells within lymphoid tissue. CD10 is also present on breast myoepithelial cells, bile canaliculi, fibroblasts, with especially high expression on the brush border of kidney and gut epithelial cells.

Type: Primary
Antigen: MME
Clonality: Monoclonal
Clone: FR4D11
Conjugation: CF405S
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR