Stampa…
Anti-SHBG Mouse Monoclonal Antibody (CF488A) [clone: SHBG/245]
Fornitore: Biotium
BNC880245-500 BNC880245-100
BNC880245-500EA 560 EUR
BNC880245-500 BNC880245-100
Anti-SHBG Mouse Monoclonal Antibody (CF488A) [clone: SHBG/245]
Anticorpi
Recognizes a protein of 45 kDa, identified as SHBG. It functions as an androgen transport protein, but may also be involved in receptor-mediated processes. Each dimer binds one molecule of steroid. It is specific for 5-alpha-dihydrotestosterone, testosterone, and 17-beta-estradiol. SHBG regulates the plasma metabolic clearance rate of steroid hormones by controlling their plasma concentration. In testis, it is synthesized by the Sertoli cells, secreted into the lumen of the seminiferous tubule and transported to the epididymis.
Type: Primary
Antigen: SHBG
Clonality: Monoclonal
Clone: SHBG/245
Conjugation: CF488A
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human