Stampa…

Anti-GFAP Mouse Monoclonal Antibody (CF594) [clone: GA-5]

Fornitore: Biotium
BNC940167-500 BNC940167-100
BNC940167-500EA 560 EUR
BNC940167-500 BNC940167-100
Anti-GFAP Mouse Monoclonal Antibody (CF594) [clone: GA-5]
Anticorpi
This MAb recognizes a protein of ~50 kDa which is identified as Glial Fibrillary Acidic Protein (GFAP). It shows no cross-reaction with other intermediate filament proteins. GFAP is specifically found in astroglia. GFAP is a very popular marker for localizing benign astrocyte and neoplastic cells of glial origin in the central nervous system. Antibody to GFAP is useful in differentiating primary gliomas from metastatic lesions in the brain and for documenting astrocytic differentiation in tumors outside the CNS.

Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: GA-5
Conjugation: CF594
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Chicken, Cow, Human, Mouse, Pig, Rabbit, Rat
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR