Stampa…

Anti-CD28 Mouse Monoclonal Antibody (Biotin) [clone: 204.12]

Fornitore: Biotium

BNCB0164-500 BNCB0164-100
BNCB0164-500EA 530 EUR
BNCB0164-500 BNCB0164-100
Anti-CD28 Mouse Monoclonal Antibody (Biotin) [clone: 204.12]
Anticorpi
Recognizes a glycoprotein of 44-88 kDa, which is identified as CD28. It is the critical T-cell co-stimulatory receptor which provides to the cell the important second activation signal by binding CD80 and CD86 that are expressed by antigen presenting cells. Besides its co-stimulation role, CD28 functions in preventing T-cells from anergic hyporesponsive state or from undergoing premature apoptotic cell death. CD28 is also expressed on human fetal NK cells and some NK cell lines, whereas on murine NK cells the CD28 expression is much broader.

Type: Primary
Antigen: CD28
Clonality: Monoclonal
Clone: 204.12
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Cow, Human, Mouse, Pig, Sheep
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR