Stampa…

Anti-MUC1 Mouse Monoclonal Antibody (CF640R) [clone: GP1.4]

Fornitore: Biotium
BNC400159-500 BNC400159-100
BNC400159-500EA 560 EUR
BNC400159-500 BNC400159-100
Anti-MUC1 Mouse Monoclonal Antibody (CF640R) [clone: GP1.4]
Anticorpi
In Western blotting, this antibody recognizes proteins in MW range of 265-400 kDa, identified as different glycoforms of EMA. This MAb reacts with the DTRP epitope in the tandem repeats. The α subunit has cell adhesive properties. It can act both as an adhesion and an anti-adhesion protein. EMA may provide a protective layer on epithelial cells against bacterial and enzyme attack. The β subunit contains a C-terminal domain, which is involved in cell signaling, through phosphorylations and protein-protein interactions. In immunohistochemical assays, it superbly stains routine formalin/paraffin carcinoma tissues. Antibody to EMA is useful as a pan-epithelial marker for detecting early metastatic loci of carcinoma in bone marrow or liver.

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: GP1.4
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human
Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR